Supelco® Organic Reference Standard, Sodium 4-hydroxybenzoate
H3766-25G
: Merck
:
- Pk:25 g
- Standards:Analytical standard
- CAS No.:114-63-6
- Description:Sodium 4-hydroxybenzoate
- Storage conditions:2...30 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00016530
- M-Clarity quality level:MQ100
- Synonym(s):Sodium p-hydroxybenzoate; 4-Hydroxybenzoic acid sodium salt
- Formula:4-HOC₆H₄CO₂Na
- Molecular weight:160,10
- GHS signal: GHS07
: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338