Supelco® Organic Reference Standard, Cysteamine S-phosphate sodium salt
Catalog # C8397-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:3724-89-8
- Description:Cysteamine S-phosphate sodium salt
- Storage conditions:2…8 °C
- Hazard class description:H302 - H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00038474
- Synonym(s):S-(2-Aminoethyl) thiophosphate monosodium salt
- Formula:C₂H₇NO₃PSNa
- Molecular weight:179,11
Specifications
About this item
- GHS signal: GHS07
Caution: P261 - P264 - P270 - P301 + P312 - P302 + P352 - P305 + P351 + P338