Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, 2-Chloro-2-deoxy-ᴅ-glucose
Supelco® Organic Reference Standard, 2-Chloro-2-deoxy-ᴅ-glucose
Catalog # C203-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 2-Chloro-2-deoxy-ᴅ-glucose
Catalog # C203-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    14685-79-1
  • Description:
    2-Chloro-2-deoxy-ᴅ-glucose
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01317996
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-CIDG
  • Melting point:
    139 - 145 °C
  • Molecular weight:
    198,60

Specifications

About this item