Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Albendazole, analytical standard, Supelco®
Albendazole, analytical standard, Supelco®
Albendazole, analytical standard, Supelco®
Catalog # A4673-10G
Supplier:
Merck
CAS Number:
Certificates
Expand image
Albendazole, analytical standard, Supelco®
Catalog # A4673-10G
Supplier:
Merck
CAS Number:
Certificates
Specifications
Pk:
10 g
Standards:
analytical standard
CAS No.:
54965-21-8
Description:
Albendazole
Description:
Albendazole
Specifications
About this item
Product Details
Documentation
Product Details
Documentation