Streptomycin solution, analytical standard, Supelco®
Catalog # 85886-10ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:~1 mg/mL in 1 mM EDTA
- Pk:10 mL
- Standards:analytical standard
- CAS No.:3810-74-0
- Description:Streptomycin solution
- M-Clarity quality level:MQ100
- Description:Streptomycin solution