Polymyxin B solution, analytical standard, Supelco®
Catalog # 81271-10ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1 mg/mL in H2O
- Pk:10 mL
- Standards:analytical standard
- CAS No.:1405-20-5
- Description:Polymyxin B solution
- M-Clarity quality level:MQ100
- Description:Polymyxin B solution