Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, C4 - C24 Even Carbon Saturated FAMEs
  49453-U
 :  Merck
 :  
Supelco® Organic Reference Standard, C4 - C24 Even Carbon Saturated FAMEs
  49453-U
 :  Merck
 :  

 

  • Concentration:
    1000 µg/ml each component in hexane
  • Pk:
    1
  • Standards:
    Analytical standard
  • Description:
    C4 - C24 Even carbon saturated FAMEs
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H304 - H315 - H336 - H361f - H373 - H411
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Flash point:
    –23 °C

 

 

  • GHS signal: GHS02, GHS08, GHS07, GHS09
 : P202 - P210 - P273 - P301 + P310 - P303 + P361 + P353 - P331