Supelco® Organic Reference Standard, F.A.M.E. Column Evaluation Mix
Catalog # 47088-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 μg/ml each component in dichloromethane
- Pk:1
- Standards:Certified reference material
- Description:F.A.M.E. Column evaluation mix
- Storage conditions:–10...–25 °C
- Hazard class description:H315 - H319 - H336 - H351
- Shipping conditions:STD
- M-Clarity quality level:MQ100
Specifications
About this item
- GHS signal: GHS08
Caution: P202 - P261 - P264 - P302 + P352 - P305 + P351 + P338 - P308 + P313