Order Entry
Slovakia
ContactUsLinkComponent
Supelco® VETRANAL® Organic Reference Standard, 4-Androstene-3,17-dione
Supelco® VETRANAL® Organic Reference Standard, 4-Androstene-3,17-dione
Catalog # 46033-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® VETRANAL® Organic Reference Standard, 4-Androstene-3,17-dione
Catalog # 46033-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    VETRANAL®, analytical standard
  • CAS No.:
    63-05-8
  • Description:
    4-Androstene-3,17-dione
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H351 - H360FD - H362
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00003615
  • M-Clarity quality level:
    MQ200
  • Synonym(s):
    Androstenedione
  • Melting point:
    170 - 171 °C (lit.)
  • Molecular weight:
    286,41

Specifications

About this item

  • GHS signal: GHS08, GHS07
Caution: P202 - P260 - P263 - P264 - P301 + P312 - P308 + P313