Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Salicylic Acid (Phenyl-¹³C₆)
  43825-5MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Salicylic Acid (Phenyl-¹³C₆)
  43825-5MG
 :  Merck
 :  

 

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    1189678-81-6
  • Description:
    Salicylic acid (phenyl-¹³C₆)
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302 - H318
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD20257120
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-Hydroxybenzoic acid (phenyl-¹³C₆)
  • Melting point:
    158...161 °C (lit.)
  • Formula:
    2-(HO)¹³C₆H₄CO₂H
  • Molecular weight:
    144,08
  • Flash point:
    157 °C - closed cup

 

 

  • GHS signal: GHS05, GHS07
 : P264 - P270 - P280 - P301 + P312 - P305 + P351 + P338 - P501