Supelco® Organic Reference Standard, Delphinidin Chloride
: Merck
- CAS No.:528-53-0
- Storage conditions:–20 °C
- Shipping conditions:STD
- MDL No.:MFCD00016663
- M-Clarity quality level:MQ100
- Synonym(s):3,3′,4′,5,5′,7-Hexahydroxyflavylium chloride; 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-1-benzopyrylium chloride
- Formula:C₁₅H₁₁ClO₇
- Molecular weight:338,70
- Storage temperature:–20 °C
- Flash point:Not applicable
Frequently Bought Together