Supelco® PESTANAL® Organic Reference Standard, Azaconazole
Catalog # 34045-100MG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:60207-31-0
- Description:Azaconazole
- Storage conditions:2...30 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00865600
- M-Clarity quality level:MQ100
- Synonym(s):1-[2-(2,4-Dichlorophenyl)-1,3-dioxolan-2-ylmethyl]-1H-1,2,4-triazole
- Molecular weight:300,14
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07
Caution: P264 - P270 - P301 + P312 - P501