Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Microcystin-LW solution, Supelco®
Microcystin-LW solution, Supelco®
Microcystin-LW solution, Supelco®
Catalog # 33763-1ML
Supplier:
Merck
CAS Number:
Expand image
Microcystin-LW solution, Supelco®
Catalog # 33763-1ML
Supplier:
Merck
CAS Number:
Specifications
Pk:
1 mL
CAS No.:
157622-02-1
Description:
Microcystin-LW solution
M-Clarity quality level:
MQ100
Description:
Microcystin-LW solution
Specifications
About this item
Product Details
Product Details