Lasalocid A sodium salt solution, analytical standard, VETRANAL™, Supelco®
Catalog # 33339-2ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 mug/mL in acetonitrile
- Pk:2 mL
- Standards:analytical standard
- CAS No.:25999-20-6
- Description:Lasalocid A sodium salt solution
- M-Clarity quality level:MQ100
- Description:Lasalocid A sodium salt solution