0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovakia
ContactUsLinkComponent
Standards
Organic Standards
Cefalonium hydrate, analytical standard, VETRANAL™, Supelco®
Cefalonium hydrate, analytical standard, VETRANAL™, Supelco®
32904-100MG
:
Merck
:
Cefalonium hydrate, analytical standard, VETRANAL™, Supelco®
32904-100MG
:
Merck
:
Pk:
100 mg
Standards:
analytical standard
CAS No.:
5575-21-3
Description:
Cefalonium hydrate
M-Clarity quality level:
MQ100
Description:
Cefalonium hydrate