Order Entry
Slovakia
ContactUsLinkComponent
Crystal Violet-d6 trihydrate, analytical standard, Supelco®
Crystal Violet-d6 trihydrate, analytical standard, Supelco®
Catalog # 32853-10MG
Supplier:  Merck
CAS Number:  
undefined
Crystal Violet-d6 trihydrate, analytical standard, Supelco®
Catalog # 32853-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    analytical standard
  • Description:
    Crystal Violet-d6 trihydrate
  • M-Clarity quality level:
    MQ100
  • Formula:
    C25D6H24ClN3 · 3H2O
  • Description:
    Crystal Violet-d6 trihydrate

Specifications

About this item