Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Fenbendazole-d3, analytical standard, VETRANAL™, Supelco®
Fenbendazole-d3, analytical standard, VETRANAL™, Supelco®
32567-10MG
:
Merck
:
Fenbendazole-d3, analytical standard, VETRANAL™, Supelco®
32567-10MG
:
Merck
:
Pk:
10 mg
Standards:
analytical standard
CAS No.:
1228182-47-5
Description:
Fenbendazole-d3
Description:
Fenbendazole-d3