0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovakia
ContactUsLinkComponent
Standards
Organic Standards
Halofuginone hydrobromide, analytical standard, VETRANAL™, Supelco®
Halofuginone hydrobromide, analytical standard, VETRANAL™, Supelco®
32481-10MG
:
Merck
:
Halofuginone hydrobromide, analytical standard, VETRANAL™, Supelco®
32481-10MG
:
Merck
:
Pk:
10 mg
Standards:
analytical standard
CAS No.:
64924-67-0
Description:
Halofuginone hydrobromide
Description:
Halofuginone hydrobromide