Nitrofurazone solution, analytical standard, VETRANAL™, Supelco®
Catalog # 31706-10ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 mug/mL in acetonitrile
- Pk:10 mL
- Standards:analytical standard
- CAS No.:59-87-0
- Description:Nitrofurazone solution
- M-Clarity quality level:MQ100
- Description:Nitrofurazone solution