Order Entry
Slovakia
ContactUsLinkComponent
Supelco® VETRANAL® Organic Reference Standard, Androsterone
Supelco® VETRANAL® Organic Reference Standard, Androsterone
Catalog # 31579-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® VETRANAL® Organic Reference Standard, Androsterone
Catalog # 31579-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    VETRANAL®, Analytical standard
  • CAS No.:
    53-41-8
  • Description:
    Androsterone
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H351 - H360FD
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00003618
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3α-Hydroxy-5α-androstan-17-one; 5α-Androstan-3α-ol-17-one
  • Melting point:
    181...184 °C (lit.)
  • Molecular weight:
    290,44

Specifications

About this item

  • GHS signal: GHS08, GHS07
Caution: P201 - P202 - P280 - P308 + P313 - P405 - P501