Erythromycin A enol ether, analytical standard, Supelco®
05238-5MG
: Merck
:
- Pk:5 mg
- Standards:analytical standard
- CAS No.:33396-29-1
- Description:Erythromycin A enol ether
- M-Clarity quality level:MQ100
- Formula:C₃₇H₆₅NO₁₂
- Description:Erythromycin A enol ether
Erythromycin A enol ether (EAEN) is an acidic degradation product of erythromycin A (EA) which is grouped under the macrolide antibiotic class of compounds.
: Bottomless glass bottle. Contents are inside inserted fused cone.