Supelco® Organic Reference Standard, C7 - C30 Saturated Alkanes
Catalog # 49451-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 µg/ml each component in hexane
- Pk:1
- Standards:Certified reference material
- Description:C7 - C30 saturated alkanes
- Storage conditions:2…8 °C
- Hazard class description:H225 - H304 - H315 - H336 - H361f - H373 - H411
- Shipping conditions:STD
- M-Clarity quality level:MQ100
Specifications
About this item
- GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P201 - P210 - P273 - P301 + P310 - P303 + P361 + P353 - P331