Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Tin-Base Solder (Lead Free)
Supelco® Organic Reference Standard, Tin-Base Solder (Lead Free)
  12242-1EA
 :  Merck
 :  
Supelco® Organic Reference Standard, Tin-Base Solder (Lead Free)
  12242-1EA
 :  Merck
 :  

 

  • Pk:
    1
  • Description:
    Tin-base solder (lead free)
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD