Organic reference standard, Zeaxanthin, analytical standard, Supelco®
Catalog # 14681-1MG-F
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mg
- Standards:Analytical standard
- CAS No.:144-68-3
- Description:Zeaxanthin, ≥95,0% (HPLC)
- M-Clarity quality level:MQ100
- Formula:C₄₀H₅₆O₂
- Storage temperature:−20 °C
- Description:Zeaxanthin
- Form:Neat
Specifications
About this item
Carotenoid pigment that contributes to the color of paprika. Zeaxanthin accumulates within the eye retina and is thought to decrease the incidence of macular degeneration.
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.