Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Vitexin 2-O-rhamnoside
  55608-25MG-F
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Vitexin 2-O-rhamnoside
  55608-25MG-F
 :  Merck
 :  

 

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    64820-99-1
  • Description:
    Vitexin 2-O-rhamnoside
  • Seq:
    01
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00143627
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-O-Rhamnosylvitexin
  • Molecular weight:
    578,52