Organic reference standard, Tutin solution, TraceCERT®, Supelco®
Catalog # 80718-0.5ML
Dodávateľ: Merck
Číslo CAS:
Špecifikácie
- Concentration:20 μg/g in acetonitrile (nominal concentration)
- Pk:0,5 mL
- CAS No.:2571-22-4
- Description:Tutin solution
- Formula:C₁₅H₁₈O₆
- Storage temperature:−20 °C
- Description:Tutin solution
Špecifikácie
O tomto produkte
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034.