Supelco® Organic Reference Standard, trans-Sinapic acid
93878-50MG
: Merck
:
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:7362-37-0
- Description:trans-Sinapic acid
- Storage conditions:2...30 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00004401
- M-Clarity quality level:MQ100
- Synonym(s):trans-4-Hydroxy-3,5-dimethoxy-cinnamic acid; trans-Sinapinic acid
- Melting point:195 - 205 °C (dec.)
- Molecular weight:224,21
- GHS signal: GHS07
: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338