Supelco® Organic Reference Standard, trans-p-Coumaric acid
Katalogové číslo 55823-50MG
Dodavatel: Merck
Číslo CAS:
Specifikace
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:501-98-4
- Description:trans-p-Coumaric acid
- Storage conditions:2...30 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00004399
- M-Clarity quality level:MQ100
- Synonym(s):p-Coumaric acid; trans-4-Hydroxycinnamic acid
- Melting point:214 °C (dec.) (lit.)
- Formula:HOC₆H₄CH=CHCO₂H
- Molecular weight:164,16
Specifikace
O tomto produktu
- GHS signal: GHS07
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338