Supelco® Organic Reference Standard, Trans-Nerolidol
18143-100MG-F
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:40716-66-3
- Description:trans-Nerolidol
- Storage conditions:2...30 °C
- Hazard class description:H317 - H319 - H410
- Shipping conditions:STD
- MDL No.:MFCD00008911
- M-Clarity quality level:MQ100
- Synonym(s):(E)-3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol; trans-3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol
- Formula:(CH₃)₂C=CHCH₂CH₂C(CH₃)=CHCH₂CH₂C(CH₃)(OH)CH=CH₂
- Molecular weight:222,37
- Density:0,876 g/ml at 25 °C (lit.)
- Flash point:113 °C - closed cup
- Boiling point:145...146 °C/12 mm Hg (lit.)
- Form:Neat
- GHS signal: GHS07
: Bottomless glass bottle. Contents are inside inserted fused cone.
: P261 - P264 - P273 - P280 - P302 + P352 - P305 + P351 + P338