Organic reference standard, Tenuazonic acid, analytical standard, Supelco®
Katalogové číslo 37018-0.1MG
Dodavatel: Merck
Číslo CAS:
Specifikace
- Pk:0,1 mg
- Standards:Analytical standard
- CAS No.:610-88-8
- Description:Tenuazonic acid
- Formula:C₁₀H₁₅NO₃
- Storage temperature:−20°C
- Description:Tenuazonic acid
- Form:Neat
Specifikace
O tomto produktu
Tenuazonic acid is a mycotoxin, which belongs to the class of tetramic acids, the largest family of natural products. It is generally isolated from phytopathogenic fungal species like Alternaria alternata, A. longipes and A. tenuissima.
Dried down, 100 μg/ml after reconstitution with 1 ml of water.