Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Tectoridin
Supelco® Organic Reference Standard, Tectoridin
  89948-50MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Tectoridin
  89948-50MG
 :  Merck
 :  

 

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    611-40-5
  • Description:
    Tectoridin
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD04039835
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    ᴅ-Glucopyranosyloxy)-5-hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one; Shekanin; Tectorigenin 7-O-β-ᴅ-glucopyranoside
  • Molecular weight:
    462,40

 

 

 : P264 - P270 - P301 + P312 - P501