Organic reference standard, T2-Toxin solution, analytical standard, Supelco®
Catalog # 34071-2ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:2 mL
- Standards:Analytical standard
- CAS No.:75-05-8
- Description:T2-Toxin solution
- M-Clarity quality level:MQ100
- Formula:C₂₄H₃₄O₉
- Description:T2-Toxin solution