Order Entry
Slovakia
ContactUsLinkComponent
Organic reference standard, Sunset Yellow FCF, analytical standard, Supelco®
Organic reference standard, Sunset Yellow FCF, analytical standard, Supelco®
Catalog # 68775-25MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Sunset Yellow FCF, analytical standard, Supelco®
Catalog # 68775-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    2783-94-0
  • Description:
    Sunset Yellow FCF, ≥95% (HPLC)
  • Storage temperature:
    −20 °C
  • Description:
    Sunset Yellow FCF
  • Form:
    Neat

Specifications

About this item

Sunset Yellow FCF azo-dye is generally used as a colorant in drug industries and in common foods.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.