Organic reference standards, Sugar Alcohol Kit, (individually packaged in quantities indicated), analytical standard, Supelco®
Catalog # 47266.
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Standards:Analytical standard
- Description:Sugar alcohol kit
- Melting point:173...175 °C
- Formula:C₂₄D₆H₁₄N₄O
- Form:Neat
Specifications
About this item
Kit components:
D-(+)Arabitol 500 mg; Dulcitol (Galactitol) 500 mg; iso-Erythritol 500 mg; Glycerol 500 mg; Maltitol 500 mg; D-Mannitol 500 mg; Ribitol (Adonitol) 500 mg; D-Sorbitol 500 mg; Xylitol 500 mg.