Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Starch from corn
Catalog # S5296-5G
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Starch from corn
Catalog # S5296-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 g
  • Standards:
    Analytical standard, Analytical standard for Starch Assay Kits SA-20 and STA-20
  • CAS No.:
    9005-25-8
  • Description:
    Starch from corn
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00082026
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Corn starch; Starch from maize
  • Formula:
    (C₆H₁₀O₅)ₙ

Specifications

About this item