Phytochemical standard, Silychristin, analytical standard, Supelco®
51681-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:33889-69-9
- Description:Silychristin
- Formula:C₂₅H₂₂O₁₀
- Storage temperature:2...8 °C
- Description:Silychristin
- Form:Neat
Silychristin is one of the congeners of flavonolignan silybin. It is usually derived from silymarin, which is extracted from the fruits of Silybum marianum (L.) Gaertn, and Asteraceae (Carduus marianus L. milk thistle).
: Bottomless glass bottle. Contents are inside inserted fused cone.