Supelco® Organic Reference Standard, Shikimic acid
Catalog # 69686-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:138-59-0
- Description:Shikimic acid
- Storage conditions:2…8 °C
- Hazard class description:H318
- Shipping conditions:WICE
- MDL No.:MFCD00066278
- M-Clarity quality level:MQ100
- Synonym(s):(3R,4S,5R)-(−)-3,4,5-Trihydroxy-1-cyclohexenecarboxylic acid
- Melting point:185 - 187 °C (lit.)
- Molecular weight:174,15
Specifications
About this item
- GHS signal: GHS05
Caution: P280 - P305 + P351 + P338