Supelco® Organic Reference Standard, Salvianolic acid A
Catalog # 97599-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:96574-01-5
- Description:Salvianolic acid A
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD08458447
- M-Clarity quality level:MQ100
- Synonym(s):(αR)-α-[[(2E)-3-[2-[(1E)-2-(3,4-Dihydroxyphenyl)ethenyl]-3,4-dihydroxyphenyl]-1-oxo-2-propen-1-yl]oxy]-3,4-dihydroxybenzenepropanoic acid
- Molecular weight:494,45