Phytochemical standard, Quercetin 3-glucuronide, analytical standard, Supelco®
90733-5MG
: Merck
:
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:22688-79-5
- Description:Quercetin 3-glucuronide, ≥90,0% (HPLC)
- Formula:C₂₁H₁₈O₁₃
- Description:Quercetin 3-glucuronide
- Form:Neat
Quercetin 3-glucuronide is a dietary flavanoid, that can be isolated from green beans.
: Bottomless glass bottle. Contents are inside inserted fused cone.