Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Quercetin 3-Glucoside
Supelco® Organic Reference Standard, Quercetin 3-Glucoside
  16654-10MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Quercetin 3-Glucoside
  16654-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    482-35-9
  • Description:
    Quercetin 3-glucoside
  • Storage conditions:
    2...8 °C
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00017746
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Quercetin 3-β-ᴅ-glucoside; 3,3′,4′,5,7-Pentahydroxyflavone 3-β-glucoside; Isoquercitrin
  • Formula:
    C₂₁H₂₀O₁₂
  • Molecular weight:
    464,38
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable
  • Form:
    Neat

 

 

 : Bottomless glass bottle. Contents are inside inserted fused cone.