Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Punicalin, mixture of anomers
Supelco® Organic Reference Standard, Punicalin, mixture of anomers
  67988-10MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Punicalin, mixture of anomers
  67988-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    65995-64-4
  • Description:
    Punicalin, mixture of anomers
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    4,6-Gallagyl-ᴅ-glucose
  • Molecular weight:
    782,53