Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Procyanidin B2
Supelco® Organic Reference Standard, Procyanidin B2
  42157-1MG-F
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Procyanidin B2
  42157-1MG-F
 :  Merck
 :  

 

  • Pk:
    1 mg
  • Standards:
    Analytical standard
  • CAS No.:
    29106-49-8
  • Description:
    Procyanidin B2
  • Storage conditions:
    2...8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01861513
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (−)-Epicatechin (4β-8)-(−)-epicatechin; 4,8″-Bi-[(+)-epicatechin]; cis,cis″-4,8″-Bi(3,3′,4′,5,7-pentahydroxyflavane); Proanthocyanidin B2
  • Formula:
    C₃₀H₂₆O₁₂
  • Molecular weight:
    578,52
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable
  • Form:
    Neat

 

 

 : Bottomless glass bottle. Contents are inside inserted fused cone.