Supelco® Organic Reference Standard, Procyanidin B1
Catalog # 19542-1MG-F
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mg
- Standards:Analytical standard
- CAS No.:20315-25-7
- Description:Procyanidin B1
- Storage conditions:2...8 °C
- Shipping conditions:STD
- MDL No.:MFCD01861512
- M-Clarity quality level:MQ100
- Synonym(s):(−)-Epicatechin (4β-8)-(+)-catechin; cis,trans′′-4,8′′-Bi-(3,3′,4′,5,7-Pentahydroxyflavane); Proanthocyanidin B1
- Formula:C₃₀H₂₆O₁₂
- Molecular weight:578,52
- Storage temperature:2...8 °C
- Flash point:Not applicable
- Form:Neat
Specifications
About this item
Packaging: 1 mg in poly bottle.