Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Procyanidin A2
Supelco® Organic Reference Standard, Procyanidin A2
Catalog # 28660-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Procyanidin A2
Catalog # 28660-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    41743-41-3
  • Description:
    Procyanidin A2
  • Storage conditions:
    2...8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD10566019
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (+)-Epicatechin-(4β-8,2β-O-7)-epicatechin; Proanthocyanidin A2
  • Formula:
    C₃₀H₂₄O₁₂
  • Molecular weight:
    576,50
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable
  • Form:
    Neat

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.