Organic reference standard, Piperine, analytical standard, Supelco®
Catalog # 75047-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:94-62-2
- Description:Piperine, ≥97,0% (HPLC)
- Melting point:131...135 °C
- Formula:C₁₇H₁₉NO₃
- Storage temperature:2...8 °C
- Description:Piperine
- Form:Neat
Specifications
About this item
Piperine is an amide isolated from the plant, Piper longum. It is a known inhibitor of hepatic and intestinal glucuronidation. Piperine is found to exhibit pharmacological properties like antitumor, antipyretic and anti-inflammatory activities.