Order Entry
Slovakia
ContactUsLinkComponent
Organic reference standard, Pigment Orange 5, analytical standard, Supelco®
Organic reference standard, Pigment Orange 5, analytical standard, Supelco®
Catalog # 09215-25MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Pigment Orange 5, analytical standard, Supelco®
Catalog # 09215-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    3468-63-1
  • Description:
    Pigment Orange 5, ≥98,0% (HPLC)
  • Melting point:
    310...320 °C
  • Formula:
    C₁₆H₁₀N₄O₅
  • Description:
    Pigment Orange 5

Specifications

About this item

Pigment orange 5 is an azo pigment, which can be produced industrially by a diazotisation and coupling sequence in which diazotised dinitroaniline is coupled into β-naphthol.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.