Supelco® Organic Reference Standard, Phospholipid mixture for HPLC from Glycine max (soybean)
Catalog # P3817-1VL
Supplier: Merck
CAS Number:
Specifications
- Pk:1 Ampoul
- Standards:Certified reference material
- Description:Phospholipid mixture for HPLC from Glycine max (soybean)
- Storage conditions:–20 °C
- Hazard class description:H302 - H315 - H319 - H331 - H336 - H351 - H361d - H372 - H412
- Shipping conditions:DICE
- M-Clarity quality level:MQ100
Specifications
About this item
- GHS signal: GHS06, GHS08
Caution: P201 - P273 - P301 + P312 + P330 - P302 + P352 - P304 + P340 + P311 - P308 + P313