Order Entry
Slovakia
ContactUsLinkComponent
Phytochemical standard, Petunidin 3-O-beta-D-glucoside chloride, analytical standard, Supelco®
Phytochemical standard, Petunidin 3-O-beta-D-glucoside chloride, analytical standard, Supelco®
Catalog # 30638-10MG
Supplier:  Merck
CAS Number:  
undefined
Phytochemical standard, Petunidin 3-O-beta-D-glucoside chloride, analytical standard, Supelco®
Catalog # 30638-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    6988-81-4
  • Description:
    Petunidin 3-O-beta-D-glucoside chloride, ≥95.0% (HPLC)
  • Formula:
    C₂₂H₂₃ClO₁₂
  • Storage temperature:
    –20 °C
  • Description:
    Petunidin 3-O-beta-D-glucoside chloride
  • Form:
    Neat

Specifications

About this item