Order Entry
Slovakia
ContactUsLinkComponent
Organic reference standard, Patulin, Supelco®
Organic reference standard, Patulin, Supelco®
Catalog # 32759-5MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Patulin, Supelco®
Catalog # 32759-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • CAS No.:
    149-29-1
  • Description:
    Patulin
  • Formula:
    C₇H₆O₄
  • Storage temperature:
    –20 °C
  • Description:
    Patulin
  • Form:
    Neat

Specifications

About this item

Patulin (PAT) is a mycotoxin of the polyketide lactone type, usually produced by species of the Penicillium, Aspergillus and Byssochlamys genera.

Fruit-based products by QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) method and solid phase extraction (SPE) followed by analysis using ultra-high performance liquid chromatography combined with electrospray ionisation tandem mass spectrometry (UHPLC-ESI-MS/MS).

PAT may be used as a reference standard for the determination of patulin in model food matrices by 'dilute and shoot' LC-MS/MS, dairy products by QuEChERS procedure and UHPLC-ESI quadrupole orbitrap (Q-Orbitrap) mass spectrometry and apple juice samples by single drop liquid-liquid-liquid microextraction (SD-LLLME) followed by LC-MS/MS analysis.