Order Entry
Slovakia
ContactUsLinkComponent
Organic reference standard, Palatinose hydrate, analytical standard, Supelco®
Organic reference standard, Palatinose hydrate, analytical standard, Supelco®
Catalog # 41556-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Palatinose hydrate, analytical standard, Supelco®
Catalog # 41556-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    343336-76-5
  • Description:
    Palatinose hydrate, ≥98,0% (HPLC)
  • Formula:
    C₁₂H₂₂O₁₁·xH₂O
  • Description:
    Palatinose hydrate

Specifications

About this item

Palatinose is a disaccharide having an α(1→6) linkage between D-glucose and D-fructose and is similar to sucrose in its physicochemical properties. It is produced from sucrose in some bacteria by the action of sucrose isomerase. In mammalian gut, palatinose is hydrolysed and absorbed much more slowly than sucrose.